Learn More
Sodium bicarbonate, 1M buffer soln., pH 8.0, Thermo Scientific Chemicals
Description
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
Specifications
Specifications
| Concentration | 1 M |
| pH | 8.0 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solubility Information | It is soluble in water. |
| IUPAC Name | sodium;hydrogen carbonate |
| Chemical Name or Material | Sodium bicarbonate buffer soln. |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only