missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Arsenite, 0.5% (w/v), Ricca Chemical
$89.10 - $170.97
Chemical Identifiers
| CAS | 7784-46-5 |
|---|---|
| Molecular Formula | AsNaO2 |
| Molecular Weight (g/mol) | 129.909 |
| InChI Key | PTLRDCMBXHILCL-UHFFFAOYSA-M |
| Synonym | sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c |
| PubChem CID | 443495 |
| ChEBI | CHEBI:29678 |
| IUPAC Name | sodium;oxoarsinite |
| SMILES | [O-][As]=O.[Na+] |
Chemical Identifiers
| 7784-46-5 | |
| 129.909 | |
| sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c | |
| CHEBI:29678 | |
| [O-][As]=O.[Na+] |
| AsNaO2 | |
| PTLRDCMBXHILCL-UHFFFAOYSA-M | |
| 443495 | |
| sodium;oxoarsinite |
Specifications
| 7784-46-5 | |
| sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c | |
| [O-][As]=O.[Na+] | |
| 129.909 | |
| CHEBI:29678 | |
| Laboratory | |
| Natural Poly Bottle | |
| Sodium Arsenite |
| AsNaO2 | |
| PTLRDCMBXHILCL-UHFFFAOYSA-M | |
| sodium;oxoarsinite | |
| 443495 | |
| 0.5% | |
| 0.5% (w/v) Aqueous Solution | |
| 500 mL |