missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenite, 0.5% (w/v), Ricca Chemical

Catalog No. 714016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
714016 500 mL Natural Poly Bottle
714032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 714016 Supplier Ricca Chemical Company Supplier No. 714016

Chemical Identifiers

CAS 7784-46-5
Molecular Formula AsNaO2
Molecular Weight (g/mol) 129.909
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
PubChem CID 443495
ChEBI CHEBI:29678
IUPAC Name sodium;oxoarsinite
SMILES [O-][As]=O.[Na+]

Specifications

Percent Purity 0.5%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.5% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material Sodium Arsenite