missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenate, Heptahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

$652.71 - $37079.56

Chemical Identifiers

CAS 10048-95-0
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613259
View Documents
Spectrum Chemical Mfg Cor
S113025GM
25 g Poly Bottle
Each for $652.71
Only null left
 
18613260
View Documents
Spectrum Chemical Mfg Cor
S1130100GM
100 g Poly Bottle
Each for $1,808.91
Only null left
 
18613261
View Documents
Spectrum Chemical Mfg Cor
S1130250GM
250 g Poly Bottle
Each for $3,175.47
Only null left
 
18613262
View Documents
Spectrum Chemical Mfg Cor
S11301KG
1 kg Poly Bottle
Each for $5,080.79
Only null left
 
18613263
View Documents
Spectrum Chemical Mfg Cor
S113010KGBL
10 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18613264
View Documents
Spectrum Chemical Mfg Cor
S113025KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10048-95-0

Specifications

10048-95-0
25 g
ACS
1
98 to 102%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.