missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenate, Heptahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1130250GM

 View more versions of this product

Catalog No. 18613261


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10048-95-0
250 g
ACS
1
98 to 102%
Poly Bottle

Chemical Identifiers

10048-95-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.