missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, For Zinc, Certified, 2.0M ±0.1M (2.0N), LabChem™
Supplier: LabChem LC228502
Specifications
| Sodium Acetate | |
| For Zinc | |
| 76,24 | |
| CH3COOHNa·3H2O | |
| 1.13g/mL | |
| Soluble in water | |
| CC(=O)[O-].O.O.O.[Na+] | |
| 136.079 | |
| CHEBI:32138 | |
| Certified | |
| Poly Bottle | |
| Traceable to NIST | |
| Fume; Carbon monoxide; Carbon dioxide | |
| Liquid |
| Colorless | |
| 6131-90-4 , 7732-18-5 | |
| C2H9NaO5 | |
| 2.0M ±0.1M (2.0N) | |
| sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
| AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
| sodium;acetate;trihydrate | |
| 23665404 | |
| 136.08 | |
| Passes Test | |
| 1.13g/mL | |
| 3.017cSt | |
| 1 L |
Chemical Identifiers
| 6131-90-4 | |
| 136.079 | |
| sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
| CHEBI:32138 | |
| CC(=O)[O-].O.O.O.[Na+] |
| C2H9NaO5 | |
| AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
| 23665404 | |
| sodium;acetate;trihydrate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature