missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, 2.00 M (2.00 N), Ricca Chemical

APHA for Zinc Testing, EPA for Fluoride Testing

Supplier:  Ricca Chemical Company 713016

 View more versions of this product

Catalog No. 7130-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Acetate
136.08 g/mol
Sodium Acetate
APHA for Zinc, EPA for Fluoride
500 mL
7732-18-5
Laboratory
Natural Poly Bottle
7.5 to 9.2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions