missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, 1.00 M (1.00 N), Ricca Chemical

Catalog No. R71295001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R71295001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R71295001A Supplier Ricca Chemical Company Supplier No. R71295001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6131-90-4 , 7732-18-5
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

CAS Min % 86.0, 11.75
CAS Max % 90.0, 12.22
Grade Laboratory
Packaging Natural Poly Bottle
pH 7.5 to 9.2
Quantity 1 L
Concentration or Composition Notes 1.00Molar,1.00Normal
Chemical Name or Material Sodium Acetate