missing translation for 'onlineSavingsMsg'
Learn More

Sodium Abietate 95.0+%, TCI America™

Catalog No. A06515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06515G 5 g
A065125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06515G Supplier TCI America Supplier No. A06515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14351-66-7
Molecular Formula C20H29NaO2
Molecular Weight (g/mol) 324.44
MDL Number MFCD00058941
InChI Key ITCAUAYQCALGGV-XTICBAGASA-M
Synonym Abietic Acid Sodium Salt
PubChem CID 133121851
IUPAC Name sodium (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylate
SMILES [Na+].CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C([O-])=O)[C@H]2CC1

Specifications

Quantity 5 g
Formula Weight 324.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Abietate