accessibility menu, dialog, popup

UserName

CAS RN 14351-66-7

IUPAC Name: sodium (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylate
Synonyms: sodium abietate
Molecular Weight (g/mol): 324.44
Molecular Formula: C20H29NaO2
InChi Key: ITCAUAYQCALGGV-XTICBAGASA-M
SMILES: [Na+].CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C([O-])=O)[C@H]2CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results