missing translation for 'onlineSavingsMsg'
Learn More

Sodium 6,7-Dihydroxynaphthalene-2-sulfonate 98.0+%, TCI America™

Catalog No. p-7126312
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059725G 25 g
D0597500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D059725G Supplier TCI America Supplier No. D059725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-53-5
Molecular Formula C10H7NaO5S
Molecular Weight (g/mol) 262.211
MDL Number MFCD00004074
InChI Key JFXDYPLHFRYDJD-UHFFFAOYSA-M
Synonym DS Acid Monosodium Salt, Monosodium 6,7-Dihydroxynaphthalene-2-sulfonate, 2,3-Dihydroxynaphthalene-6-sulfonic Acid Monosodium Salt
PubChem CID 4410831
IUPAC Name sodium;6,7-dihydroxynaphthalene-2-sulfonate
SMILES C1=CC(=CC2=CC(=C(C=C21)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 262.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 6,7-Dihydroxynaphthalene-2-sulfonate
EINECSNumber (4)-0511
TSCA Yes