accessibility menu, dialog, popup

UserName

CAS RN 135-53-5

NaHOHOSOOO

CAS RN 135-53-5

IUPAC Name: sodium 6,7-dihydroxynaphthalene-2-sulfonate
Synonyms: sodium 6,7-dihydroxynaphthalene-2-sulfonate
Molecular Weight (g/mol): 262.21
Molecular Formula: C10H7NaO5S
InChi Key: JFXDYPLHFRYDJD-UHFFFAOYSA-M
SMILES: [Na+].OC1=C(O)C=C2C=C(C=CC2=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)