missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-(tert-butylcarbonyloxy)benzenesulfonate, 95%

Catalog No. p-7057130
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6649606 5 g
AAH6649614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6649606 Supplier Thermo Scientific Chemicals Supplier No. H6649606
Only null left

CAS: 188114-91-2 | C11H13NaO5S | 280.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188114-91-2
Molecular Formula C11H13NaO5S
Molecular Weight (g/mol) 280.27
MDL Number MFCD09859793
InChI Key YJWRXNUCTLGRAC-UHFFFAOYSA-M
Synonym sodium 4-pivaloyloxy benzenesulfonate, sodium 4-tert-butylcarbonyloxy-benzensulfonate, sodium 4-t-butylcarbonyloxy-benzensulfonate, sodium 4-t-butylcarbonyloxy benzensulfonate, sodium 4-2,2-dimethylpropanoyl oxy benzenesulfonate, sodium 4-t-butylcarbonyloxy-benzensulf..., 4-pivaloyloxy benzenesulfonic acid sodium salt, 2,2-dimethylpropanoic acid 4-sulfophenyl ester sodium salt, sodium 4-2,2-dimethylpropanoyl oxy benzene-1-sulfonate, propanoic acid, 2,2-dimethyl-, 4-sulfophenyl ester, sodium salt 1:1
PubChem CID 23717480
IUPAC Name sodium;4-(2,2-dimethylpropanoyloxy)benzenesulfonate
SMILES CC(C)(C)C(=O)OC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 5 g
Formula Weight 280.27
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Sodium 4-(tert-butylcarbonyloxy)benzenesulfonate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only