accessibility menu, dialog, popup

UserName

CAS RN 188114-91-2

IUPAC Name: sodium 4-[(2,2-dimethylpropanoyl)oxy]benzene-1-sulfonate
Synonyms: sodium 4-[(2,2-dimethylpropanoyl)oxy]benzenesulfonate
Molecular Weight (g/mol): 280.27
Molecular Formula: C11H13NaO5S
InChi Key: YJWRXNUCTLGRAC-UHFFFAOYSA-M
SMILES: [Na+].CC(C)(C)C(=O)OC1=CC=C(C=C1)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results