missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Hydroxy-3-methoxybenzoate 98.0+%, TCI America™

Catalog No. H026525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H026525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H026525G Supplier TCI America Supplier No. H026525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28508-48-7
Molecular Formula C8H7NaO4
Molecular Weight (g/mol) 190.13
MDL Number MFCD00016534
InChI Key ZFRVFUWCVYLUIH-UHFFFAOYSA-M
Synonym 4-Hydroxy-3-methoxybenzoic Acid Sodium Salt, Sodium Vanillate, Vanillic Acid Sodium Salt
PubChem CID 23675709
IUPAC Name sodium;4-hydroxy-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)[O-])O.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 190.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Hydroxy-3-methoxybenzoate