missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 4-Chlorobenzenesulfinate 98.0+%, TCI America™
Supplier: TCI America C155925G
Specifications
| Sodium 4-Chlorobenzenesulfinate | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD00035602 | |
| JFXAUUFCZJYLJF-UHFFFAOYSA-M | |
| sodium 4-chlorobenzene-1-sulfinate | |
| 23664783 | |
| ≥98.0% (T) |
| 14752-66-0 | |
| 25 g | |
| C6H4ClNaO2S | |
| sodium 4-chlorobenzenesulfinate, 4-chlorobenzenesulfinic acid sodium salt, sodium 4-chlorobenzene sulfinate, unii-x2u129fz4n, sodium p-chlorobenzenesulphinate, sodium 4-chlorobenzene-1-sulfinate, 4-chlorobenzene sulfinic acid sodium, 4-chlorobenzenesulfinic acid sodium salt hydrate, p-chlorobenzenesulfinic acid, sodium salt, acmc-1cin1 | |
| [Na+].[O-]S(=O)C1=CC=C(Cl)C=C1 | |
| 198.60 | |
| 198.60 |
Chemical Identifiers
| 14752-66-0 | |
| 198.60 | |
| JFXAUUFCZJYLJF-UHFFFAOYSA-M | |
| 23664783 | |
| [Na+].[O-]S(=O)C1=CC=C(Cl)C=C1 |
| C6H4ClNaO2S | |
| MFCD00035602 | |
| sodium 4-chlorobenzenesulfinate, 4-chlorobenzenesulfinic acid sodium salt, sodium 4-chlorobenzene sulfinate, unii-x2u129fz4n, sodium p-chlorobenzenesulphinate, sodium 4-chlorobenzene-1-sulfinate, 4-chlorobenzene sulfinic acid sodium, 4-chlorobenzenesulfinic acid sodium salt hydrate, p-chlorobenzenesulfinic acid, sodium salt, acmc-1cin1 | |
| sodium 4-chlorobenzene-1-sulfinate |
Safety and Handling
TSCA : No