accessibility menu, dialog, popup

UserName

CAS RN 14752-66-0

IUPAC Name: sodium 4-chlorobenzene-1-sulfinate
Synonyms: sodium 4-chlorobenzenesulfinate
Molecular Weight (g/mol): 198.6
Molecular Formula: C6H4ClNaO2S
InChi Key: JFXAUUFCZJYLJF-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)