missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Chlorobenzenesulfinate 98.0+%, TCI America™

Catalog No. p-7125060
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C15595G 5 g
C155925G 25 g
2 options

Catalog No. C15595G

Supplier: TCI America C15595G

Only null left

Chemical Identifiers

CAS 14752-66-0
Molecular Formula C6H4ClNaO2S
Molecular Weight (g/mol) 198.60
MDL Number MFCD00035602
InChI Key JFXAUUFCZJYLJF-UHFFFAOYSA-M
Synonym sodium 4-chlorobenzenesulfinate, 4-chlorobenzenesulfinic acid sodium salt, sodium 4-chlorobenzene sulfinate, unii-x2u129fz4n, sodium p-chlorobenzenesulphinate, sodium 4-chlorobenzene-1-sulfinate, 4-chlorobenzene sulfinic acid sodium, 4-chlorobenzenesulfinic acid sodium salt hydrate, p-chlorobenzenesulfinic acid, sodium salt, acmc-1cin1
PubChem CID 23664783
IUPAC Name sodium 4-chlorobenzene-1-sulfinate
SMILES [Na+].[O-]S(=O)C1=CC=C(Cl)C=C1

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 198.60
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium 4-Chlorobenzenesulfinate