missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 4-Aminoazobenzene-4'-sulfonate 98.0+%, TCI America™
Supplier: TCI America A074825G
Specifications
| Sodium 4-Aminoazobenzene-4′-sulfonate | |
| C12H10N3NaO3S | |
| 25 g | |
| FIXVWFINKCQNFG-UHFFFAOYSA-M | |
| sodium;4-[(4-aminophenyl)diazenyl]benzenesulfonate | |
| 23691997 | |
| ≥98.0% (HPLC) |
| 2491-71-6 | |
| MFCD00035564 | |
| yellow rfs, 11648 yellow, acid yellow fwa, new yellow gmf, 4-aminoazobenzene-4'-sulfonic acid sodium salt, hexacol yellow rfs, c.i. food yellow 6, monosodium salt, sodium 4'-aminoazobenzene-4-sulphonate, yellow r.i.s., unii-z452e7h2u8 | |
| C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
| 299.28 | |
| 299.28 | |
| Crystalline Powder |
Chemical Identifiers
| 2491-71-6 | |
| 299.28 | |
| FIXVWFINKCQNFG-UHFFFAOYSA-M | |
| 23691997 | |
| C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
| C12H10N3NaO3S | |
| MFCD00035564 | |
| yellow rfs, 11648 yellow, acid yellow fwa, new yellow gmf, 4-aminoazobenzene-4'-sulfonic acid sodium salt, hexacol yellow rfs, c.i. food yellow 6, monosodium salt, sodium 4'-aminoazobenzene-4-sulphonate, yellow r.i.s., unii-z452e7h2u8 | |
| sodium;4-[(4-aminophenyl)diazenyl]benzenesulfonate |
Safety and Handling
EINECSNumber : (3)-2001
TSCA : Yes