missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Aminoazobenzene-4'-sulfonate 98.0+%, TCI America™

Catalog No. p-7117042
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074825G 25 g
A0748500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A074825G Supplier TCI America Supplier No. A074825G
Only null left

Chemical Identifiers

CAS 2491-71-6
Molecular Formula C12H10N3NaO3S
Molecular Weight (g/mol) 299.28
MDL Number MFCD00035564
InChI Key FIXVWFINKCQNFG-UHFFFAOYSA-M
Synonym yellow rfs, 11648 yellow, acid yellow fwa, new yellow gmf, 4-aminoazobenzene-4'-sulfonic acid sodium salt, hexacol yellow rfs, c.i. food yellow 6, monosodium salt, sodium 4'-aminoazobenzene-4-sulphonate, yellow r.i.s., unii-z452e7h2u8
PubChem CID 23691997
IUPAC Name sodium;4-[(4-aminophenyl)diazenyl]benzenesulfonate
SMILES C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 299.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Aminoazobenzene-4′-sulfonate
EINECSNumber (3)-2001
TSCA Yes