accessibility menu, dialog, popup

UserName

CAS RN 2491-71-6

IUPAC Name: sodium 4-[2-(4-aminophenyl)diazen-1-yl]benzene-1-sulfonate
Synonyms: sodium 4-[2-(4-aminophenyl)diazen-1-yl]benzenesulfonate
Molecular Weight (g/mol): 299.28
Molecular Formula: C12H10N3NaO3S
InChi Key: FIXVWFINKCQNFG-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)