missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Nitrosalicylate 99.0+%, TCI America™

Catalog No. p-7133710
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02585G 5 g
N025825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02585G Supplier TCI America Supplier No. N02585G
Only null left

Chemical Identifiers

CAS 164915-53-1
Molecular Formula C7H4NNaO5
Molecular Weight (g/mol) 205.10
MDL Number MFCD00070529
InChI Key DLXOJVOMIITILS-UHFFFAOYSA-M
Synonym 3-Nitrosalicylic Acid Sodium Salt
PubChem CID 54726778
IUPAC Name sodium 2-carboxy-6-nitrobenzen-1-olate
SMILES [Na+].OC(=O)C1=C([O-])C(=CC=C1)[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 205.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Nitrosalicylate