accessibility menu, dialog, popup

UserName

NOOOOHONa

CAS RN 164915-53-1

IUPAC Name: sodium 2-carboxy-6-nitrobenzen-1-olate
Synonyms: sodium 2-carboxy-6-nitrobenzenolate
Molecular Weight (g/mol): 205.1
Molecular Formula: C7H4NNaO5
InChi Key: DLXOJVOMIITILS-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)C1=C([O-])C(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results