missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-nitrobenzoate, 94%, may contain up to ca 10% water

Catalog No. AAL1002836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1002836 500 g
AAL1002822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1002836 Supplier Thermo Scientific Chemicals Supplier No. L1002836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 827-95-2 | C7H4NNaO4 | 189.10 g/mol

Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 827-95-2
Molecular Formula C7H4NNaO4
Molecular Weight (g/mol) 189.10
MDL Number MFCD00150535,MFCD00051097
InChI Key MUADFEZFSKAZLT-UHFFFAOYSA-M
Synonym sodium 3-nitrobenzoate, 3-nitrobenzoic acid sodium salt, benzoic acid, 3-nitro-, sodium salt, unii-i1eq837b3j, m-nitrobenzoic acid sodium salt, sodium m-nitrobenzoate, benzoic acid, 3-nitro-, sodium salt 1:1, 3-nitrobenzoic acid, sodium salt, acmc-209pqe, dsstox_cid_24733
PubChem CID 70014
IUPAC Name sodium;3-nitrobenzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point >300°C
Quantity 500 g
Beilstein 4166479
Solubility Information Soluble in water.
Formula Weight 189.1
Percent Purity 94%
Assay may contain up to ≈10% water
Chemical Name or Material Sodium 3-nitrobenzoate
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 212-578-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only