CAS RN 827-95-2
IUPAC Name:
sodium 3-nitrobenzoate
Synonyms:
sodium 3-nitrobenzoate
Molecular Weight (g/mol):
189.1
Molecular Formula:
C7H4NNaO4
InChi Key:
MUADFEZFSKAZLT-UHFFFAOYSA-M
SMILES:
[Na+].[O-]C(=O)C1=CC=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Sodium 3-nitrobenzoate, 94%, may contain up to ca 10% water
Sodium 3-nitrobenzoate, 94%, may contain up to ca 10% water
CAS: 827-95-2 Molecular Formula: C7H4NNaO4 Molecular Weight (g/mol): 189.10 MDL Number: MFCD00150535,MFCD00051097 InChI Key: MUADFEZFSKAZLT-UHFFFAOYSA-M Synonym: sodium 3-nitrobenzoate,3-nitrobenzoic acid sodium salt,benzoic acid, 3-nitro-, sodium salt,unii-i1eq837b3j,m-nitrobenzoic acid sodium salt,sodium m-nitrobenzoate,benzoic acid, 3-nitro-, sodium salt 1:1,3-nitrobenzoic acid, sodium salt,acmc-209pqe,dsstox_cid_24733 PubChem CID: 70014 IUPAC Name: sodium;3-nitrobenzoate SMILES: [Na+].[O-]C(=O)C1=CC=CC(=C1)[N+]([O-])=O