missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonate 98.0+%, TCI America™

Catalog No. p-7135948
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0826200MG 200 mg
S08261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0826200MG Supplier TCI America Supplier No. S0826200MG
Only null left

Chemical Identifiers

CAS 82692-96-4
Molecular Formula C12H22NNaO7S
Molecular Weight (g/mol) 347.36
MDL Number MFCD09261431
InChI Key PNGLEYLFMHGIQO-UHFFFAOYNA-M
Synonym 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonic Acid Sodium Salt, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3-methoxyaniline Sodium Salt, ADOS
PubChem CID 71307074
IUPAC Name sodium 3-[ethyl(3-methoxyphenyl)amino]-2-hydroxypropane-1-sulfonate dihydrate
SMILES O.O.[Na+].CCN(CC(O)CS([O-])(=O)=O)C1=CC(OC)=CC=C1

Specifications

Melting Point 144°C
Color White
Quantity 200 mg
Formula Weight 311.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonate [for Biochemical Research]