accessibility menu, dialog, popup

UserName

CH3NSOOOOHOCH3H2OH2ONa

CAS RN 82692-96-4

IUPAC Name: sodium 3-[ethyl(3-methoxyphenyl)amino]-2-hydroxypropane-1-sulfonate dihydrate
Synonyms: sodium 3-[ethyl(3-methoxyphenyl)amino]-2-hydroxypropane-1-sulfonate dihydrate
Molecular Weight (g/mol): 347.36
Molecular Formula: C12H22NNaO7S
InChi Key: PNGLEYLFMHGIQO-UHFFFAOYNA-M
SMILES: O.O.[Na+].CCN(CC(O)CS([O-])(=O)=O)C1=CC(OC)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results