missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Methylsalicylate 97.0+%, TCI America™

Catalog No. C041125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C041125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C041125G Supplier TCI America Supplier No. C041125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32768-20-0
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.131
MDL Number MFCD00058985
InChI Key ODAXNLPYGWAGTH-UHFFFAOYSA-M
Synonym o-Cresotic Acid Sodium Salt, 2-Hydroxy-3-methylbenzoic Acid Sodium Salt, 2-Hydroxy-m-toluic Acid Sodium Salt, Sodium 2-Hydroxy-3-methylbenzoate, 3-Methylsalicylic Acid Sodium Salt
PubChem CID 23668291
IUPAC Name sodium;2-hydroxy-3-methylbenzoate
SMILES CC1=CC=CC(=C1O)C(=O)[O-].[Na+]

Specifications

Color Red-White
Quantity 25 g
Formula Weight 174.13
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Methylsalicylate