accessibility menu, dialog, popup

UserName

NaCH3OOHO

CAS RN 32768-20-0

IUPAC Name: sodium 2-hydroxy-3-methylbenzoate
Synonyms: sodium 2-hydroxy-3-methylbenzoate
Molecular Weight (g/mol): 174.13
Molecular Formula: C8H7NaO3
InChi Key: ODAXNLPYGWAGTH-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=CC(C([O-])=O)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results