missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Hydroxy-2-naphthoate 98.0+%, TCI America™

Catalog No. p-7130462
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02845G 5 g
H028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02845G Supplier TCI America Supplier No. H02845G
Only null left

Chemical Identifiers

CAS 14206-62-3
Molecular Formula C11H7NaO3
Molecular Weight (g/mol) 210.164
MDL Number MFCD00068251
InChI Key ZCNGWQCRXUGQCW-UHFFFAOYSA-M
Synonym 3-Hydroxy-2-naphthoic Acid Sodium Salt, 2-Hydroxy-3-naphthoic Acid Sodium Salt
PubChem CID 23672297
IUPAC Name sodium;3-hydroxynaphthalene-2-carboxylate
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)[O-])O.[Na+]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 210.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Hydroxy-2-naphthoate
EINECSNumber (4)-0399
TSCA Yes