accessibility menu, dialog, popup

UserName

NaHOOO

CAS RN 14206-62-3

IUPAC Name: sodium 3-hydroxynaphthalene-2-carboxylate
Synonyms: sodium 3-hydroxy-2-naphthoate
Molecular Weight (g/mol): 210.16
Molecular Formula: C11H7NaO3
InChi Key: ZCNGWQCRXUGQCW-UHFFFAOYSA-M
SMILES: [Na+].OC1=C(C=C2C=CC=CC2=C1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results