missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-[Ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonate Hydrate 98.0+%, TCI America™

Catalog No. p-7135939
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08051G 1 g
S08055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08051G Supplier TCI America Supplier No. S08051G
Only null left

Chemical Identifiers

CAS 82692-93-1
Molecular Formula C12H18NNaO4S
Molecular Weight (g/mol) 295.329
MDL Number MFCD00040641
InChI Key IRQRBVOQGUPTLG-UHFFFAOYSA-M
Synonym toos, 3-ethyl m-tolyl amino-2-hydroxypropane-1-sulfonic acid, sodium salt, sodium 3-n-ethyl-3-methylanilino-2-hydroxypropanesulfonate, sodium n-ethyl-n-2-hydroxy-3-sulfopropyl-m-toluidine, sodium 3-ethyl m-tolyl amino-2-hydroxypropane-1-sulfonate, sodium 3-ethyl 3-methylphenyl amino-2-hydroxypropane-1-sulfonate, 3-n-ethyl-3-methylanilino-2-hydroxypropanesulfonic acid sodium salt, 1-propanesulfonic acid, 3-ethyl 3-methylphenyl amino-2-hydroxy-, monosodium salt, 3-ethyl 3-methylphenyl amino-2-hydroxy-1-propanesulfonic acid monosodium salt, n-ethyl-n-2-hydroxy-3-sulfopropyl-m-toluidine sodium salt
PubChem CID 23676159
IUPAC Name sodium;3-(N-ethyl-3-methylanilino)-2-hydroxypropane-1-sulfonate
SMILES CCN(CC(CS(=O)(=O)[O-])O)C1=CC=CC(=C1)C.[Na+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 295.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-[Ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonate Hydrate