accessibility menu, dialog, popup

UserName

CH3NSOOOOHH3CNa

CAS RN 82692-93-1

IUPAC Name: sodium 3-[ethyl(3-methylphenyl)amino]-2-hydroxypropane-1-sulfonate
Synonyms: sodium 3-[ethyl(3-methylphenyl)amino]-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol): 295.33
Molecular Formula: C12H18NNaO4S
InChi Key: IRQRBVOQGUPTLG-UHFFFAOYNA-M
SMILES: [Na+].CCN(CC(O)CS([O-])(=O)=O)C1=CC=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results