missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-Naphthoate 98.0+%, TCI America™
Supplier: TCI America N061725G
Specifications
| Sodium 2-Naphthoate | |
| White | |
| MFCD00671570 | |
| 2-Naphthalenecarboxylic Acid Sodium Salt, 2-Naphthoic Acid Sodium Salt | |
| [Na+].[O-]C(=O)C1=CC2=CC=CC=C2C=C1 | |
| 194.17 | |
| 194.16 | |
| Crystalline Powder |
| 17273-79-9 | |
| C11H7NaO2 | |
| 25 g | |
| WUJXROKYJRAVIO-UHFFFAOYSA-M | |
| sodium naphthalene-2-carboxylate | |
| 23685274 | |
| ≥98.0% (T) |
Chemical Identifiers
| 17273-79-9 | |
| 194.17 | |
| WUJXROKYJRAVIO-UHFFFAOYSA-M | |
| 23685274 | |
| [Na+].[O-]C(=O)C1=CC2=CC=CC=C2C=C1 |
| C11H7NaO2 | |
| MFCD00671570 | |
| 2-Naphthalenecarboxylic Acid Sodium Salt, 2-Naphthoic Acid Sodium Salt | |
| sodium naphthalene-2-carboxylate |
Safety and Handling
EINECSNumber : (3)-1272
TSCA : No