missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Naphthoate 98.0+%, TCI America™

Catalog No. p-7133895
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06175G 5 g
N061725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06175G Supplier TCI America Supplier No. N06175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17273-79-9
Molecular Formula C11H7NaO2
Molecular Weight (g/mol) 194.17
MDL Number MFCD00671570
InChI Key WUJXROKYJRAVIO-UHFFFAOYSA-M
Synonym 2-Naphthalenecarboxylic Acid Sodium Salt, 2-Naphthoic Acid Sodium Salt
PubChem CID 23685274
IUPAC Name sodium naphthalene-2-carboxylate
SMILES [Na+].[O-]C(=O)C1=CC2=CC=CC=C2C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 194.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Naphthoate
EINECSNumber (3)-1272
TSCA No