missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-Naphthalenesulfonate 98.0+%, TCI America™
Supplier: TCI America N001625G
Specifications
| Sodium 2-Naphthalenesulfonate | |
| White-Yellow | |
| MFCD00064186 | |
| sodium 2-naphthalenesulfonate, sodium naphthalene-2-sulfonate, 2-naphthalenesulfonic acid sodium salt, 2-naphthalenesulfonic acid, sodium salt, unii-r5f0ctd2oj, sodium-2-naphthalenesulfonate, sodium naphthalene-6-sulfonate, 2-naphthalene sulfonic acid sodium salt, sodium beta-naphthalenesulfonate, sodium naphthalene-2-sulphonate | |
| [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1 | |
| 230.21 | |
| 230.21 | |
| Crystalline Powder |
| 532-02-5 | |
| C10H7NaO3S | |
| 25 g | |
| YWPOLRBWRRKLMW-UHFFFAOYSA-M | |
| sodium naphthalene-2-sulfonate | |
| 23661868 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 532-02-5 | |
| 230.21 | |
| YWPOLRBWRRKLMW-UHFFFAOYSA-M | |
| 23661868 | |
| [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1 |
| C10H7NaO3S | |
| MFCD00064186 | |
| sodium 2-naphthalenesulfonate, sodium naphthalene-2-sulfonate, 2-naphthalenesulfonic acid sodium salt, 2-naphthalenesulfonic acid, sodium salt, unii-r5f0ctd2oj, sodium-2-naphthalenesulfonate, sodium naphthalene-6-sulfonate, 2-naphthalene sulfonic acid sodium salt, sodium beta-naphthalenesulfonate, sodium naphthalene-2-sulphonate | |
| sodium naphthalene-2-sulfonate |
Safety and Handling
EINECSNumber : (4)-0466
RTECSNumber : QK3678000
TSCA : Yes