accessibility menu, dialog, popup

UserName

NaOSOO

CAS RN 532-02-5

IUPAC Name: sodium naphthalene-2-sulfonate
Synonyms: sodium 2-naphthalenesulfonate
Molecular Weight (g/mol): 230.21
Molecular Formula: C10H7NaO3S
InChi Key: YWPOLRBWRRKLMW-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (4)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (6)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)