missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Naphthalenesulfonate 98.0+%, TCI America™

Catalog No. p-7131360
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03535G 5 g
I035325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03535G Supplier TCI America Supplier No. I03535G
Only null left

Chemical Identifiers

CAS 532-02-5
Molecular Formula C10H7NaO3S
Molecular Weight (g/mol) 230.21
MDL Number MFCD00064186
InChI Key YWPOLRBWRRKLMW-UHFFFAOYSA-M
Synonym sodium 2-naphthalenesulfonate, sodium naphthalene-2-sulfonate, 2-naphthalenesulfonic acid sodium salt, 2-naphthalenesulfonic acid, sodium salt, unii-r5f0ctd2oj, sodium-2-naphthalenesulfonate, sodium naphthalene-6-sulfonate, 2-naphthalene sulfonic acid sodium salt, sodium beta-naphthalenesulfonate, sodium naphthalene-2-sulphonate
PubChem CID 23661868
IUPAC Name sodium naphthalene-2-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1

Specifications

Color White
Quantity 5 g
Formula Weight 230.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Naphthalenesulfonate
EINECSNumber (4)-0466
RTECSNumber QK3678000
TSCA Yes