missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Iodobenzenesulfonate 97.0+%, TCI America™

Catalog No. S08481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08481G 1 g
S08485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08481G Supplier TCI America Supplier No. S08481G

Chemical Identifiers

CAS 62973-69-7
Molecular Formula C6H4INaO3S
Molecular Weight (g/mol) 306.049
MDL Number MFCD12031329
InChI Key KIPWXZMYZCPXGE-UHFFFAOYSA-M
Synonym 2-Iodobenzenesulfonic Acid Sodium Salt
PubChem CID 23662047
IUPAC Name sodium;2-iodobenzenesulfonate
SMILES C1=CC=C(C(=C1)S(=O)(=O)[O-])I.[Na+]

Specifications

Quantity 1 g
Formula Weight 306.05
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Iodobenzenesulfonate