accessibility menu, dialog, popup

UserName

SOOOINa

CAS RN 62973-69-7

IUPAC Name: sodium 2-iodobenzene-1-sulfonate
Synonyms: sodium 2-iodobenzenesulfonate
Molecular Weight (g/mol): 306.05
Molecular Formula: C6H4INaO3S
InChi Key: KIPWXZMYZCPXGE-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results