missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Chlorotoluene-4-sulfonate 95.0+%, TCI America™

Catalog No. C0298500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0298500G 500 g
C029825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0298500G Supplier TCI America Supplier No. C0298500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5138-91-0
Molecular Formula C7H6ClNaO3S
Molecular Weight (g/mol) 228.622
MDL Number MFCD00053619
InChI Key FQFDFCOTXFCZMB-UHFFFAOYSA-M
Synonym 2-Chlorotoluene-4-sulfonic Acid Sodium Salt, Sodium 3-Chloro-4-methylbenzenesulfonate
PubChem CID 23678886
IUPAC Name sodium;3-chloro-4-methylbenzenesulfonate
SMILES CC1=C(C=C(C=C1)S(=O)(=O)[O-])Cl.[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 228.62
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Chlorotoluene-4-sulfonate