accessibility menu, dialog, popup

UserName

NaCH3ClSOHOO

CAS RN 5138-91-0

IUPAC Name: 3-chloro-4-methylbenzene-1-sulfonic acid sodium
Synonyms: 3-chloro-4-methylbenzenesulfonic acid sodium
Molecular Weight (g/mol): 229.63
Molecular Formula: C7H7ClNaO3S
InChi Key: FCGGMSDOURZDTN-UHFFFAOYSA-N
SMILES: [Na].CC1=CC=C(C=C1Cl)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results