missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonate 95.0+%, TCI America™

Catalog No. p-7127063
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20115G 5 g
D201125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20115G Supplier TCI America Supplier No. D20115G
Only null left

Chemical Identifiers

CAS 54495-25-9
Molecular Formula C7H2F3N2NaO7S
Molecular Weight (g/mol) 338.145
MDL Number MFCD00059068
InChI Key PYGGEZQHHQFWCY-UHFFFAOYSA-M
Synonym 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonic Acid Sodium Salt
PubChem CID 2737132
IUPAC Name sodium;2,6-dinitro-4-(trifluoromethyl)benzenesulfonate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])S(=O)(=O)[O-])[N+](=O)[O-])C(F)(F)F.[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 338.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonate