accessibility menu, dialog, popup

UserName

NOOSOOONOOFFFNa

CAS RN 54495-25-9

IUPAC Name: sodium 2,6-dinitro-4-(trifluoromethyl)benzene-1-sulfonate
Synonyms: sodium 2,6-dinitro-4-(trifluoromethyl)benzenesulfonate
Molecular Weight (g/mol): 338.14
Molecular Formula: C7H2F3N2NaO7S
InChi Key: PYGGEZQHHQFWCY-UHFFFAOYSA-M
SMILES: [Na+].[O-][N+](=O)C1=CC(=CC(=C1S([O-])(=O)=O)[N+]([O-])=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results