missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonate 95.0+%, TCI America™

Catalog No. d20115g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D20115G 5 g
D201125G 25 g
2 options

Catalog No. D20115G

Supplier: TCI America D20115G

Only null left

Chemical Identifiers

CAS 54495-25-9
Molecular Formula C7H2F3N2NaO7S
Molecular Weight (g/mol) 338.145
MDL Number MFCD00059068
InChI Key PYGGEZQHHQFWCY-UHFFFAOYSA-M
Synonym 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonic Acid Sodium Salt
PubChem CID 2737132
IUPAC Name sodium;2,6-dinitro-4-(trifluoromethyl)benzenesulfonate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])S(=O)(=O)[O-])[N+](=O)[O-])C(F)(F)F.[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 338.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonate