missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,4-Dimethylbenzenesulfonate Monohydrate 98.0+%, TCI America™

Catalog No. D068225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D068225G 25 g
D0682500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D068225G Supplier TCI America Supplier No. D068225G

Chemical Identifiers

CAS 142063-30-7
Molecular Formula C8H11NaO4S
Molecular Weight (g/mol) 226.222
MDL Number MFCD00041883
InChI Key LAOKFSDVOMOLJF-UHFFFAOYSA-M
Synonym 2,4-Dimethylbenzenesulfonic Acid Sodium Salt, Sodium m-Xylene-4-sulfonate, m-Xylene-4-sulfonic Acid Sodium Salt
PubChem CID 70700101
IUPAC Name sodium;2,4-dimethylbenzenesulfonate;hydrate
SMILES CC1=CC(=C(C=C1)S(=O)(=O)[O-])C.O.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,4-Dimethylbenzenesulfonate Monohydrate
EINECSNumber (3)-1909
TSCA Yes