accessibility menu, dialog, popup

UserName

CH3SOOOCH3H2ONa

CAS RN 142063-30-7

IUPAC Name: sodium 2,4-dimethylbenzene-1-sulfonate hydrate
Synonyms: sodium 2,4-dimethylbenzenesulfonate hydrate
Molecular Weight (g/mol): 226.22
Molecular Formula: C8H11NaO4S
InChi Key: LAOKFSDVOMOLJF-UHFFFAOYSA-M
SMILES: O.[Na+].CC1=CC(C)=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results