missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-Propanesulfonate 98.0+%, TCI America™
Supplier: TCI America I03415G
Specifications
| Sodium 1-Propanesulfonate [Reagent for Ion-Pair Chromatography] | |
| 255°C | |
| C3H7NaO3S | |
| 5 g | |
| NPAWNPCNZAPTKA-UHFFFAOYSA-M | |
| sodium;propane-1-sulfonate | |
| 4319363 | |
| ≥98.0% (T) |
| 14533-63-2 | |
| White | |
| MFCD00062546 | |
| sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt | |
| CCCS(=O)(=O)[O-].[Na+] | |
| 146.136 | |
| 146.14 | |
| Crystalline Powder |
Chemical Identifiers
| 14533-63-2 | |
| 146.136 | |
| NPAWNPCNZAPTKA-UHFFFAOYSA-M | |
| 4319363 | |
| CCCS(=O)(=O)[O-].[Na+] |
| C3H7NaO3S | |
| MFCD00062546 | |
| sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt | |
| sodium;propane-1-sulfonate |
Safety and Handling
TSCA : No