missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Propanesulfonate 98.0+%, TCI America™

Catalog No. p-7131348
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I03415G 5 g
I034125G 25 g
2 options

Catalog No. I03415G

Supplier: TCI America I03415G

Only null left

Chemical Identifiers

CAS 14533-63-2
Molecular Formula C3H7NaO3S
Molecular Weight (g/mol) 146.136
MDL Number MFCD00062546
InChI Key NPAWNPCNZAPTKA-UHFFFAOYSA-M
Synonym sodium 1-propanesulfonate, sodium propanesulphonate, 1-propanesulfonic acid sodium salt, sodium propane-1-sulfonate, sodium propanesulfonate, acmc-209cuo, ipc-alks-3, c3h7o3s.na, potassium propane-1-sulfonate, propanesulfonic acid sodium salt
PubChem CID 4319363
IUPAC Name sodium;propane-1-sulfonate
SMILES CCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 255°C
Color White
Quantity 5 g
Formula Weight 146.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Propanesulfonate [Reagent for Ion-Pair Chromatography]