accessibility menu, dialog, popup

UserName

NaCH3SOOO

CAS RN 14533-63-2

IUPAC Name: sodium propane-1-sulfonate
Synonyms: sodium propane-1-sulfonate
Molecular Weight (g/mol): 146.14
Molecular Formula: C3H7NaO3S
InChi Key: NPAWNPCNZAPTKA-UHFFFAOYSA-M
SMILES: [Na+].CCCS([O-])(=O)=O
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)